Bovine;Horse;Human;Rat Thymosin beta₄ Acetate
Supplier: BACHEM AMERICAS INC
4043020.0001 4043020.0005
H-2608.0001BAEA 1290.81 CAD
H-2608.0001BA H-2608.0005BA
Bovine;Horse;Human;Rat Thymosin beta₄ Acetate
Proteins and Peptides
Thymosin β₄ is a 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide. Extracellular thymosin β₄ may contribute to physiological processes such as angiogenesis, wound healing, and regulation of inflammation. Additionally to its numerous functions thymosin β₄ might also be of therapeutic value in the setting of acute myocardial damage.