(beta-Asp⁵)-Delta-Sleep Inducing Peptide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4028635.0005 4028635.0025
H-2545.0005BAEA 1161.77 CAD
H-2545.0005BA H-2545.0025BA
(beta-Asp⁵)-Delta-Sleep Inducing Peptide Trifluoroacetate
Proteins and Peptides
This DSIP analog serves as a good substrate for protein L-isoaspartyl methyltransferase, a n enzyme occuring in the brain involved in the repair of age-damaged proteins containing atypical iosaspartyl peptide bonds.