Toxic Shock Syndrome Toxin-1 (TSST-1) (58-78)
Supplier: BACHEM AMERICAS INC
4031315.0500
4031315.1000
H-2522.0500BAEA
1464.03
CAD
H-2522.0500BA
H-2522.1000BA
Toxic Shock Syndrome Toxin-1 (TSST-1) (58-78)
Proteins and Peptides
TSST-1 is a "superantigen" which binds to MHC class II molecules and induces stimulation of T cells. This TSST-1 fragment has an active site of the TSST-1 holotoxin which binds to MHC class II molecules and is able to xenostimulate T cells in an MHC unrestricted manner. Therefore this peptide could be of great value in studying the immunobiology of MHC class II molecules.