Ac-Asp-Glu-Val-Asp-aldehyde (Pseudo Acid)
Supplier: BACHEM AMERICAS INC
4026022.0005
4026022.0025
H-2496.0005BAEA
565.21
CAD
H-2496.0005BA
H-2496.0025BA
Ac-Asp-Glu-Val-Asp-aldehyde (Pseudo Acid)
Proteins and Peptides
Ac-DEVD-CHO is a potent inhibitor of the poly(ADP-ribose) polymerase (PARP) cleavage by caspase-3 (IC₅₀ = 0.2 nM). PARP cleavage occurs at the onset of apoptosis. The peptide aldehyde was able to attenuate apoptotic events in vitro (IC₅₀ = 10 to 100 nM).