Bovine;Dog;Human;Mouse;Rat (Ser¹⁴⁰)-Myelin Proteolipid Protein (139-151) (depalmitoylated) Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4030575.0001 4030575.0005
H-2478.0001BAEA 387.21 CAD
H-2478.0001BA H-2478.0005BA
Bovine;Dog;Human;Mouse;Rat (Ser¹⁴⁰)-Myelin Proteolipid Protein (139-151) (depalmitoylated) Trifluoroacetate
Proteins and Peptides

This fragment of myelin proteolipid protein is an encephalitogenic determinant, able to bind to diverse sets of T cell receptors.

Order Now

SPECIFICATIONS

Protein/peptide name (Ser¹⁴⁰)-Myelin Proteolipid Protein (139-151) (depalmitoylated) Trifluoroacetate
Environmentally Preferable
Protein synonyms (Ser140)-Lipophilin (139-151) (depalmitoylated)|(Ser140)-PLP (139-151) (depalmitoylated)
Species Human,Bovine,Dog,Rat,Mouse
Source
CAS No. 122018-58-0
MDL No. MFCD18782147
Sequence H-His-Ser-Leu-Gly-Lys-Trp-Leu-Gly-His-Pro-Asp-Lys-Phe-OH
Molecular weight 1521.74
Molecular formula C₇₂H₁₀₄N₂₀O₁₇
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR