Bovine beta-Casomorphin
Supplier: BACHEM AMERICAS INC
4003340.0005 4003340.0025
H-2280.0005BAEA 206.51 CAD
H-2280.0005BA H-2280.0025BA
Bovine beta-Casomorphin
Proteins and Peptides
Besides deltorphins, dermorphins, dynorphins, enkephalins and endorphins, Bachem offers opioid peptides such as acetalins, BAM peptides, α-casein and gluten exorphins, endomorphins, kyotorphins, metorphamide, opiorphins, δ opioid receptor antagonists, and valorphin.
  • Also called β-casein exorphins.
BCM-7, YPFPGPI, has first been isolated from an enzymatic casein digest. Only cow casein variants A1 and B yield this bioactive peptide after in vitro digestion with intestinal enzymes, but not the common A2 variant. β-Casomorphin displays opioid properties. The β-casomorphin generated by digestion of bovine milk could play a role in the aetiology of human diseases as type 1 diabetes and cardiovascular affections. In a rat model, BCM-7 attenuated the development of nephropathy in type I diabetes via inhibition of epithelial-mesenchymal transition of renal tubular epithelial cells.
A hypothalamic opioid peptide related to Leu-enkephalin and dynorphin A with potent activity in the guinea-pig ileum assay.