Human Calcitonin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4014409.0001 4014409.0005
H-2250.0001BAEA 440.12 CAD
H-2250.0001BA H-2250.0005BA
Human Calcitonin Trifluoroacetate
Proteins and Peptides
For Elcatonin see H-2214.
Calcitonin is a peptide hormone involved in calcium and phosphorus homeostasis. Its inhibitory effect on osteoclast activity lowers blood calcium levels and positively influences bone mass density.