Glutathione-diethyl ester (reduced)
Supplier: BACHEM AMERICAS INC
4025802.0250
4025802.1000
H-2238.0250BAEA
1414.1
CAD
H-2238.0250BA
H-2238.1000BA
Glutathione-diethyl ester (reduced)
Proteins and Peptides
Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.
Human cells transport glutathione diethyl ester much more effectively than the corresponding monoethyl ester (H-1298). Hence, the diethyl ester of glutathione could be used for decreasing oxidative stress and toxicity.