Eel (Des-Cys¹,cyclo(Ser²-Asu⁷))-Calcitonin Acetate
Supplier: BACHEM AMERICAS INC
4025810.0001
4025810.0005
H-2214.0001BAEA
516.34
CAD
H-2214.0001BA
H-2214.0005BA
Eel (Des-Cys¹,cyclo(Ser²-Asu⁷))-Calcitonin Acetate
Proteins and Peptides
For Elcatonin see H-2214.
Elcatonin, also known as carbocalcitonin, is the aminosuberic acid analog of eel calcitonin. It has all the biological properties of the corresponding natural calcitonin (H-2255). The substitution of the disulfide bond of natural calcitonins with an ethylene bridge in 1-7 N-terminal position gives elcatonin greater stability and excellent tolerability when used in vivo.