H-D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH₂
Supplier: BACHEM AMERICAS INC
4025623.0001 4025623.0005
H-2186.0001BAEA 206.51 CAD
H-2186.0001BA H-2186.0005BA
H-D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH₂
Proteins and Peptides
The somatostatin analog CTOP is a very potent and highly selective ligand for µ-opioid receptors. CTOP shows only little interaction with other opioid receptor systems.