(D-2-Nal⁵,Cys⁶·¹¹,Tyr⁷,D-Trp⁸,Val¹⁰,2-Nal¹²)-Somatostatin-14 (5-12) amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4025467.0001 4025467.0005
H-2126.0001BAEA 206.44 CAD
H-2126.0001BA H-2126.0005BA
(D-2-Nal⁵,Cys⁶·¹¹,Tyr⁷,D-Trp⁸,Val¹⁰,2-Nal¹²)-Somatostatin-14 (5-12) amide Trifluoroacetate
Proteins and Peptides
BIM-23042 functions as neuromedin B receptor antagonist (IC₅₀ = 885 ± 98 nM). It displays a > 100-fold higher selectivity for neuromedin B versus gastrin-releasing peptide receptors.