Human (DL-Isoser¹)-TRAP-6 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4025390.0001
4025390.0005
H-1944.0001BAEA
522.6
CAD
H-1944.0001BA
H-1944.0005BA
Human (DL-Isoser¹)-TRAP-6 Trifluoroacetate
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
PAR-1 peptides are also called TRAP peptides (thrombin receptor-activating peptides).
This racemic isoserine analog of SFLLRN (H-8365) has only 15-20% of the platelet aggregating activity of SFLLRN. In compensation iso-SFLLRN significantly resists degradation by aminopeptidase M and is much more stable in plasma and serum.