(D-Arg¹,D-Trp⁷·⁹,Leu¹¹)-Substance P
Supplier: BACHEM AMERICAS INC
4004691.0001 4004691.0005 4004691.0025
H-1925.0001BAEA 206.51 CAD
H-1925.0001BA H-1925.0005BA H-1925.0025BA
(D-Arg¹,D-Trp⁷·⁹,Leu¹¹)-Substance P
Proteins and Peptides
Spantide I, a high potency substance P antagonist with a weak spasmogenic and a poor histamine releasing effect.