H-Arg-Gly-Asp-OH, Bachem
Supplier: BACHEM AMERICAS INC
4009173.0025 4009173.0100
H-1830.0025BAEA 309.81 CAD
H-1830.0025BA H-1830.0100BA
H-Arg-Gly-Asp-OH, Bachem
Proteins and Peptides
The target epitope of several integrin receptors is the RGD sequence, a cell adhesion motif shared by several matrix-associated adhesive glycoproteins, such as fibronectin, vitronectin, fibrinogen, collagen type I, laminin, and von Willebrand factor.