(Arg⁸)-Vasopressin (free acid) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4005475.0005 4005475.0025
H-1775.0005BAEA 619.42 CAD
H-1775.0005BA H-1775.0025BA
(Arg⁸)-Vasopressin (free acid) Trifluoroacetate
Proteins and Peptides
For desmopressin see H-7675.
Compared to AVP, AVP (free acid) shows markedly reduced antidiuretic activity and almost no vasopressor activity in vivo.