Angiotensin II Acetate
Supplier: BACHEM AMERICAS INC
4006473.0025 4006473.0100
H-1705.0025BAEA 324.91 CAD
H-1705.0025BA H-1705.0100BA
Angiotensin II Acetate
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
Angiotensin II, DRVYIHPF, is the major biologically active component of the renin-angiotensin system. Its activities include the regulation of blood pressure, salt and water balance, and vascular tonus. Ang II stimulates various enzymes such as NADPH oxidase. Ang II is often used as a model compound in the development and evaluation of analytical methods for peptides and proteins (e.g. MS).