Gluten Exorphin A5

Supplier: BACHEM AMERICAS INC

4025054.0025 4025054.0100
H-1668.0025BAEA 1161.77 CAD
H-1668.0025BA H-1668.0100BA
Gluten Exorphin A5
Proteins and Peptides

Besides deltorphins, dermorphins, dynorphins, enkephalins and endorphins, Bachem offers opioid peptides such as acetalins, BAM peptides, α-casein and gluten exorphins, endomorphins, kyotorphins, metorphamide, opiorphins, δ opioid receptor antagonists, and valorphin.


The sequence of this opioid peptide was found at 15 sites in the primary structure of the high molecular weight glutenin. Gluten exorphin A5 is highly specific for δ-receptors and displays opioid activity in the MVD assay. The N-terminal Gly increases its activity - thus the structure-activity relationships of gluten exorphins A are quite different from those of the endogenous opioid peptides.

Order Now

SPECIFICATIONS

Protein/peptide name Gluten Exorphin A5
Environmentally Preferable
Species
Source
CAS No. 142155-24-6
MDL No. MFCD00237075
Sequence H-Gly-Tyr-Tyr-Pro-Thr-OH
Molecular weight 599.64
Molecular formula C₂₉H₃₇N₅O₉
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR