H-Ala-His-Lys-OH Acetate, Bachem
Supplier: BACHEM AMERICAS INC
4012005.0250 4012005.1000
H-1555.0250BAEA 877.75 CAD
H-1555.0250BA H-1555.1000BA
H-Ala-His-Lys-OH Acetate, Bachem
Proteins and Peptides
The tripeptide AHK, a GHK analog, also forms complexes with Cu(II). AHK-Cu promotes the growth of human hair follicles in vitro.