Human beta-MSH Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4030885.0001 4030885.0005
H-1475.0001BAEA 645.43 CAD
H-1475.0001BA H-1475.0005BA
Human beta-MSH Trifluoroacetate
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name beta-MSH Trifluoroacetate
Environmentally Preferable
Species Human
Source
CAS No. 17908-57-5
MDL No. MFCD04113546
Sequence H-Ala-Glu-Lys-Lys-Asp-Glu-Gly-Pro-Tyr-Arg-Met-Glu-His-Phe-Arg-Trp-Gly-Ser-Pro-Pro-Lys-Asp-OH
Molecular weight 2660.95
Molecular formula C₁₁₈H₁₇₄N₃₄O₃₅S
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR