Human IL-6 (88-121)
Supplier: BACHEM AMERICAS INC
4025280.0500 4025280.1000
H-1398.0500BAEA 1059.11 CAD
H-1398.0500BA H-1398.1000BA
Human IL-6 (88-121)
Proteins and Peptides
Interleukins are soluble proteins that act as local hormones. They possess powerful immunoregulatory properties.
IL-6 (88-121) competitively inhibits the binding of human IL-6 to its receptor by interacting with the IL-6 receptor to a significant degree. It is suggested that this peptide comprises a part of the receptor-binding region of IL-6.