Glutathione-monoethyl ester (reduced)
Supplier: BACHEM AMERICAS INC
4015775.0100 4015775.0250
H-1298.0100BAEA 344.23 CAD
H-1298.0100BA H-1298.0250BA
Glutathione-monoethyl ester (reduced)
Proteins and Peptides
Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.
Glutathione-monoethyl ester is effectively transported into many cell types including erythrocytes and converted intracellularly into glutathione. The compound may therefore be useful as a protective agent against the effects of radiation and other types of toxicity as well as in experiments on glutathione transport, metabolism, and function.