H-Ala-Ala-Ala-Ala-Ala-Ala-OH, Bachem
Supplier: BACHEM AMERICAS INC
4010634.0250 4010634.1000
H-1280.0250BAEA 1448.37 CAD
H-1280.0250BA H-1280.1000BA
H-Ala-Ala-Ala-Ala-Ala-Ala-OH, Bachem
Proteins and Peptides
Oligomers of L- and/or D-alanine, the trimers to the hexamers. Bachem also offers terminally modified oligoalanines as H-1005, Ac-Ala₄-OH. and H-1010, Ac-Ala₄-OMe.
Hexaalanine.