H-Ala-Ala-Ala-Ala-Ala-OH, Bachem
Supplier: BACHEM AMERICAS INC
4007939.0100 4007939.0250
H-1270.0100BAEA 258.14 CAD
H-1270.0100BA H-1270.0250BA
H-Ala-Ala-Ala-Ala-Ala-OH, Bachem
Proteins and Peptides
Oligomers of L- and/or D-alanine, the trimers to the hexamers. Bachem also offers terminally modified oligoalanines as H-1005, Ac-Ala₄-OH. and H-1010, Ac-Ala₄-OMe.
Pentaalanine with its strong tendency to form β-sheets may serve as a model for amyloid fibrillation and interactions of small molecules with Aβ.