H-Ala-Ala-Ala-Ala-OH, Bachem
Supplier: BACHEM AMERICAS INC
4000024.0250
4000024.1000
H-1260.0250BAEA
476.62
CAD
H-1260.0250BA
H-1260.1000BA
H-Ala-Ala-Ala-Ala-OH, Bachem
Proteins and Peptides
Oligomers of L- and/or D-alanine, the trimers to the hexamers. Bachem also offers terminally modified oligoalanines as H-1005, Ac-Ala₄-OH. and H-1010, Ac-Ala₄-OMe.
Tetraalanine is a substrate for carboxypeptidase P from pig kidneys.