Indolicidin
Supplier: BACHEM AMERICAS INC
4030762.0500 4030762.1000
H-1234.0500BAEA 1007.25 CAD
H-1234.0500BA H-1234.1000BA
Indolicidin
Proteins and Peptides
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
This cationic, tryptophan-rich peptide was purified from bovine granulocytes. Indolicidin shows potent antimicrobial activity in vitro against bacteria and fungi. It is also highly cytotoxic to T-line cells.