Human;Mouse H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-NH₂
Supplier: BACHEM AMERICAS INC
4017117.0001
4017117.0005
H-1224.0001BAEA
387.21
CAD
H-1224.0001BA
H-1224.0005BA
Human;Mouse H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-NH₂
Proteins and Peptides
The peptide CDPGYIGSR amide, which comprises residues 925-933 of the laminin B1 chain, inhibited both angiogenesis and solid tumor growth.