N¹-Glutathionyl-spermidine disulfide
Supplier: BACHEM AMERICAS INC
4015761.0005 4015761.0025
H-1126.0005BAEA 903.37 CAD
H-1126.0005BA H-1126.0025BA
N¹-Glutathionyl-spermidine disulfide
Proteins and Peptides
Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.
The enzyme trypanothione reductase from the insect trypanosomatid Crithidia fasciculata not only uses trypanothione as a substrate, but also this related acyclic disulfide (Km = 149 µM).