(Nle⁴,D-Phe⁷)-alpha-MSH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4003800.0001 4003800.0005
H-1100.0001BAEA 278.74 CAD
H-1100.0001BA H-1100.0005BA
(Nle⁴,D-Phe⁷)-alpha-MSH Trifluoroacetate
Proteins and Peptides
Afamelanotide, a synthetic analog of α-MSH which shows prolonged biological activity and is 26 times more active than α-MSH in the adenylate cyclase assay.