RF9 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4061350.0005 4061350.0025
G-4800.0005BAEA 516.34 CAD
G-4800.0005BA G-4800.0025BA
RF9 Trifluoroacetate
Proteins and Peptides
Potent and selective neuropeptide FF (NPFF) receptor antagonist. RF9 (1-Adamantanecarbonyl-Arg-Phe-NH₂) blocked the delayed and long-lasting paradoxical opioid-induced hyperalgesia and prevented the development of associated tolerance. RF9 could also efficiently block the increase in blood pressure and heart rate evoked by NPFF.