H-Asp(Phe-OH)-OH, Bachem
Supplier: BACHEM AMERICAS INC
4033064.0250 4033064.1000
G-4750.0250BAEA 570.65 CAD
G-4750.0250BA G-4750.1000BA
H-Asp(Phe-OH)-OH, Bachem
Proteins and Peptides
β-Asp-Phe was isolated and identified in urine and plasma from healthy human volunteers. It has also been used in studies investigating structure-taste relationships of dipeptides.