Cyclo(-Met-Pro)
Supplier: BACHEM AMERICAS INC
4029457.0250 4029457.1000
G-4725.0250BAEA 570.65 CAD
G-4725.0250BA G-4725.1000BA
Cyclo(-Met-Pro)
Proteins and Peptides
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
Cyclo(Met-Pro) could be detected in grilled and stewed beef. The cyclopeptide seems to be an important component of beef flavor.