Dansyl-Ala-Arg-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4028360.0005
4028360.0025
G-4600.0005BAEA
794.35
CAD
G-4600.0005BA
G-4600.0025BA
Dansyl-Ala-Arg-OH Trifluoroacetate
Proteins and Peptides
Fluorescent substrate for carboxypeptidase M. Because the substrate and the cleavage product Dansyl-Ala-OH are equally fluorescent (λex = 340 nm; λem = 495 nm), the product has to be extracted with chloroform. The uncleaved substrate remains in the aqueous phase (at acidic pH).