Pyr-Trp-OH
Supplier: BACHEM AMERICAS INC
4027664.0250 4027664.1000
G-4545.0250BAEA 697.9 CAD
G-4545.0250BA G-4545.1000BA
Pyr-Trp-OH
Proteins and Peptides
These dipeptides may potentially serve as drug carriers to facilitate diffusion across the blood-brain barrier. Both peptides well permeate through gastro-intestinal lipid membranes, the acidic molecule permeating slower than the ester derivative. As their peptide bonds are stable, both dipeptides are suitable for oral administration.