H-Gly-Gly-OEt · HCl
Supplier: BACHEM AMERICAS INC
4018277.0025 4018277.0100
G-4220.0025BAEA 476.62 CAD
G-4220.0025BA G-4220.0100BA
H-Gly-Gly-OEt · HCl
Proteins and Peptides
Used as a low molecular model compound for the nitrosation of peptides and proteins.