H-D-Phe-D-Phe-OH
Supplier: BACHEM AMERICAS INC
4008206.0250 4008206.1000
G-3805.0250BAEA 464.74 CAD
G-3805.0250BA G-3805.1000BA
H-D-Phe-D-Phe-OH
Proteins and Peptides
D-Phe-D-Phe forms enzymatically stable nanotubes from HFIP solution.