H-Cys-Gly-OH
Supplier: BACHEM AMERICAS INC
4002969.0250 4002969.1000
G-3755.0250BAEA 387.21 CAD
G-3755.0250BA G-3755.1000BA
H-Cys-Gly-OH
Proteins and Peptides
Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.
This dipeptide found in plasma and urine is generated during the catabolism of glutathione. Cysteinylglycine has been suggested to induce oxidative stress and lipid peroxidation, leading to the development of human cancers. For the oxidized form see G-1815.