H-Val-Ala-OH
Supplier: BACHEM AMERICAS INC
4007094.0001 4007094.0005
G-3500.0001BAEA 697.9 CAD
G-3500.0001BA G-3500.0005BA
H-Val-Ala-OH
Proteins and Peptides
A hydrophobic nanotube-forming dipeptide. The pores absorb and store gases as methane or hydrogen.