H-Tyr-Leu-OH
Supplier: BACHEM AMERICAS INC
4002549.0001 4002549.0005
G-3430.0001BAEA 529.57 CAD
G-3430.0001BA G-3430.0005BA
H-Tyr-Leu-OH
Proteins and Peptides
These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.
A dipeptide from β-lactoglobulin showing ACE-inhibitory activity, IC₅₀ 122.1 μmol.