H-Pro-Lys-OH Acetate

Supplier: BACHEM AMERICAS INC

4000520.0250 4000520.1000
G-3040.0250BAEA 413.58 CAD
G-3040.0250BA G-3040.1000BA
H-Pro-Lys-OH Acetate
Proteins and Peptides

Substrate for prolinase (prolyl dipeptidase).

Order Now

SPECIFICATIONS

Protein/peptide name H-Pro-Lys-OH Acetate
Environmentally Preferable
Species
Source
CAS No. 71227-70-8
MDL No. MFCD02259544
Sequence H-Pro-Lys-OH
Molecular weight 243.31
Molecular formula C₁₁H₂₁N₃O₃
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR