H-Pro-Arg-OH Acetate
Supplier: BACHEM AMERICAS INC
4006116.0250 4006116.1000
G-2990.0250BAEA 464.74 CAD
G-2990.0250BA G-2990.1000BA
H-Pro-Arg-OH Acetate
Proteins and Peptides
These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.
PR inhibited angiotensin-1 converting enzyme (ACE), IC₅₀ 4.1 μM.