H-Pro-Ala-OH
Supplier: BACHEM AMERICAS INC
4011264.0001
4011264.0005
G-2985.0001BAEA
413.58
CAD
G-2985.0001BA
G-2985.0005BA
H-Pro-Ala-OH
Proteins and Peptides
Substrate for human kidney prolinase (prolyl dipeptidase), used for monitoring the enzyme purification.