H-Phe-Phe-OH
Supplier: BACHEM AMERICAS INC
4000605.0001
4000605.0005
G-2925.0001BAEA
238.26
CAD
G-2925.0001BA
G-2925.0005BA
H-Phe-Phe-OH
Proteins and Peptides
When evaporating HFIP solutions of diphenylalanine, nanotubes are formed. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation.