µ-Conotoxin GIIIB
Supplier: BACHEM AMERICAS INC
4016344.0500 4016344.1000
H-9015.0500BAEA 1588.74 CAD
H-9015.0500BA H-9015.1000BA
µ-Conotoxin GIIIB
Proteins and Peptides
This strongly basic µ-conotoxin from the venom of Conus geographus is highly effective in blocking sodium channels in vertebrate skeletal muscle, thereby preventing the propagation of muscle action potentials.