H-Gly-Gly-Tyr-OH
Supplier: BACHEM AMERICAS INC
4007920.0001 4007920.0005
H-3495.0001BAEA 529.57 CAD
H-3495.0001BA H-3495.0005BA
H-Gly-Gly-Tyr-OH
Proteins and Peptides
These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.
The tripeptide GGY showed ACE-inhibitory activity, IC₅₀ 1.3 µmol/l.