H-Leu-Trp-OH
Supplier: BACHEM AMERICAS INC
4030830.0001 4030830.0005
G-2605.0001BAEA 877.75 CAD
G-2605.0001BA G-2605.0005BA
H-Leu-Trp-OH
Proteins and Peptides
These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.
LW is an ACE-inhibitory dipeptide found in fermented milk, IC₅₀ 6.6 μM. Substrate for aminopeptidase W, Km 0.57 mM and kcat 6770 min⁻¹.