H-Leu-Leu-OMe · HBr
Supplier: BACHEM AMERICAS INC
4000725.0001
4000725.0005
G-2550.0001BAEA
329.6
CAD
G-2550.0001BA
G-2550.0005BA
H-Leu-Leu-OMe · HBr
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
The lysosomotropic dipeptide LLME has selective toxicity for NK cells, CTL, and M phi without adverse effects on a variety of other lymphoid or nonlymphoid cell types. See also E-2080.