Kyotorphin acetate
Supplier: BACHEM AMERICAS INC
4003049.0250 4003049.1000
G-2450.0250BAEA 439.41 CAD
G-2450.0250BA G-2450.1000BA
Kyotorphin acetate
Proteins and Peptides
Besides deltorphins, dermorphins, dynorphins, enkephalins and endorphins, Bachem offers opioid peptides such as acetalins, BAM peptides, α-casein and gluten exorphins, endomorphins, kyotorphins, metorphamide, opiorphins, δ opioid receptor antagonists, and valorphin.
The analgesic, opioid dipeptide kyotorphin was first identified in bovine brain. Administered intracisternally to mice, kyotorphin had an analgesic effect which was about 4.2 times more potent than that of Met-enkephalin. Kyotorphin modulates the Na⁺/Cl⁻dependant opioid peptide transporter.