H-Ile-Tyr-OH
Supplier: BACHEM AMERICAS INC
4002273.0001 4002273.0005
G-2440.0001BAEA 516.34 CAD
G-2440.0001BA G-2440.0005BA
H-Ile-Tyr-OH
Proteins and Peptides
These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.
Potent dipeptide angiotensin I-converting enzyme (ACE) inhibitor, IC₅₀ 0.008 mg/ml. Application of IY reduced blood pressure in spontaneously hypertensive rats.