H-His-Ser-OH
Supplier: BACHEM AMERICAS INC
4008023.0250 4008023.1000
G-2340.0250BAEA 258.14 CAD
G-2340.0250BA G-2340.1000BA
H-His-Ser-OH
Proteins and Peptides
The 1:1 complex of histidylserine with Co(II)(Cys-Gly) was shown to cleave DNA by hydrolysis at physiological pH. Cleavage has also been observed employing the analogous Co complex with His-Phe (N-1095).