H-His-Lys-OH · HBr
Supplier: BACHEM AMERICAS INC
4007285.0250 4007285.1000
G-2320.0250BAEA 464.74 CAD
G-2320.0250BA G-2320.1000BA
H-His-Lys-OH · HBr
Proteins and Peptides
A metabolite of GHK (H-3510) observed in rat plasma.